siRNA Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297625.1 | NP_001284554.1 | 3, 4 | 271 | ||
| NM_144684.3 | NP_653285.2 | 4, 5 | 398 | |||
| XM_011526465.2 | XP_011524767.1 | 4, 5 | 800 | |||
| GenBank mRNA | AK096559.1 | - | 4, 5 | 388 | ||
| AL832560.1 | - | 3, 4 | 248 | |||
| AY512662.1 | - | 4, 5 | 367 | |||
| BC007056.1 | - | 4, 5 | 393 | |||
| BC018783.1 | - | 4, 5 | 367 | 4873910 | ||
| BC054018.1 | - | 4, 5 | 398 | |||
| BC065503.1 | - | 4, 5 | 373 | |||
| DA501214.1 | - | 4, 5 | 396 | |||

ZNF480
Genomic Map