siRNA Details
| Gene Symbol: CCDC155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144688.4 | NP_653289.3 | 10, 11 | 1077 | ||
| XM_005258549.3 | XP_005258606.1 | 10, 11 | 1077 | |||
| XM_011526488.2 | XP_011524790.1 | 10, 11 | 1077 | |||
| XM_011526489.2 | XP_011524791.1 | 10, 11 | 2977 | |||
| XM_011526490.2 | XP_011524792.1 | 10, 11 | 1077 | |||
| XM_011526494.2 | XP_011524796.1 | 11, 12 | 1050 | |||
| XM_011526495.2 | XP_011524797.1 | 11, 12 | 1086 | |||
| XM_011526496.2 | XP_011524798.1 | 9, 10 | 938 | |||
| XM_011526497.2 | XP_011524799.1 | 10, 11 | 1036 | |||
| XM_017026333.1 | XP_016881822.1 | 9, 10 | 1210 | |||
| GenBank mRNA | AK057220.1 | - | 10, 11 | 1046 | ||
| BC029811.1 | - | 10, 11 | 996 | 5171712 | ||

CCDC155
Genomic Map