siRNA Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300838.1 | NP_001287767.1 | 2 | 361 | ||
| NM_001300839.1 | NP_001287768.1 | 2 | 107 | |||
| NM_001300841.1 | NP_001287770.1 | 2 | 123 | |||
| NM_144697.3 | NP_653298.1 | 1 | 950 | |||
| XM_017000378.1 | XP_016855867.1 | 2 | 292 | |||
| GenBank mRNA | AK056960.1 | - | 1 | 231 | ||
| AK098755.1 | - | 1 | 634 | |||
| AK315397.1 | - | 1 | 635 | |||
| BC017397.1 | - | 2 | 79 | |||
| BC027999.1 | - | 2 | 351 | 5194413 | ||
| BC047238.1 | - | 1 | 664 | 5273646 | ||
| BE784098.1 | - | 2 | 53 | |||

CIART
Genomic Map