siRNA Details
| Gene Symbol: SMCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144775.2 | NP_658988.2 | 1 | 884 | ||
| GenBank mRNA | AF467440.1 | - | 1 | 762 | ||
| AK296847.1 | - | 1 | 872 | |||
| BC001018.2 | - | 1 | 762 | |||
| BC005067.1 | - | 1 | 762 | |||
| BC014179.2 | - | 1 | 762 | |||
| BC101115.1 | - | 1 | 758 | |||
| BC101116.2 | - | 1 | 758 | |||
| BC101117.2 | - | 1 | 758 | |||
| BC142680.1 | - | 1 | 854 | 40148405 | ||

SMCR8
Genomic Map