siRNA Details
| Gene Symbol: CDH24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022478.3 | NP_071923.2 | 3 | 533 | ||
| NM_144985.3 | NP_659422.2 | 3 | 533 | |||
| XM_011537089.1 | XP_011535391.1 | 3 | 614 | |||
| GenBank mRNA | AK057922.1 | - | 3 | 469 | ||
| AK289576.1 | - | 3 | 533 | |||
| AK307746.1 | - | 3 | 515 | |||
| AY260900.1 | - | 2 | 366 | |||
| AY260901.1 | - | 2 | 366 | |||
| AY358199.1 | - | 3 | 489 | |||
| BC110867.1 | - | 3 | 504 | |||
| BX248750.1 | - | 3 | 515 | |||

CDH24
Genomic Map