siRNA Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144994.7 | NP_659431.5 | 6 | 599 | ||
| GenBank mRNA | AF492401.1 | - | 6 | 575 | ||
| AJ315766.1 | - | 5 | 543 | |||
| AK057011.1 | - | 6 | 599 | |||
| AK092564.1 | - | 5 | 473 | |||
| AY196212.1 | - | 6 | 583 | |||
| BC107056.2 | - | 6 | 594 | 40005436 | ||

ANKRD23
Genomic Map