siRNA Details
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198963.1 | NP_945314.1 | 4, 5 | 679 | ||
| XM_011533154.2 | XP_011531456.1 | 4, 5 | 683 | |||
| XM_011533155.2 | XP_011531457.1 | 4, 5 | 681 | |||
| XM_011533156.2 | XP_011531458.1 | 4, 5 | 683 | |||
| XM_011533157.1 | XP_011531459.1 | 4, 5 | 683 | |||
| GenBank mRNA | AK057423.1 | - | 3, 4 | 479 | ||
| AK296739.1 | - | 4, 5 | 718 | |||
| AK307514.1 | - | 4, 5 | 708 | |||
| BC039270.1 | - | 3, 4 | 486 | |||
| BC060778.1 | - | 4, 5 | 679 | |||
| BC131534.1 | - | 4, 5 | 651 | 40082400 | ||

DHX57
Genomic Map