siRNA Details
| Gene Symbol: ZNF582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320371.1 | NP_001307300.1 | 5 | 2456 | ||
| NM_144690.2 | NP_653291.1 | 5 | 2234 | |||
| GenBank mRNA | AK055489.1 | - | 5 | 2153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF582| Gene Symbol: ZNF582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320371.1 | NP_001307300.1 | 5 | 2456 | ||
| NM_144690.2 | NP_653291.1 | 5 | 2234 | |||
| GenBank mRNA | AK055489.1 | - | 5 | 2153 | ||