siRNA Details
| Gene Symbol: ZNF558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304350.1 | NP_001291279.1 | 4, 5 | 925 | ||
| NM_144693.2 | NP_653294.1 | 9, 10 | 1111 | |||
| XM_011527703.2 | XP_011526005.1 | 5, 6 | 612 | |||
| XM_017026370.1 | XP_016881859.1 | 10, 11 | 1305 | |||
| XM_017026371.1 | XP_016881860.1 | 11, 12 | 1427 | |||
| XM_017026372.1 | XP_016881861.1 | 11, 12 | 1538 | |||
| GenBank mRNA | AK055494.1 | - | 5, 6 | 653 | ||
| AK291265.1 | - | 9, 10 | 1066 | |||
| AK301664.1 | - | 4, 5 | 693 | |||
| BC064356.1 | - | 5, 6 | 489 | 5115196 | ||

ZNF558
Genomic Map