siRNA Details
| Gene Symbol: AIFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018060.2 | NP_001018070.1 | 9 | 988 | ||
| NM_001146288.1 | NP_001139760.1 | 9 | 1073 | |||
| NM_144704.2 | NP_653305.1 | 9 | 988 | |||
| NR_027464.1 | 8 | 945 | ||||
| GenBank mRNA | AK055035.1 | - | 9 | 972 | ||
| AK094844.1 | - | 9 | 977 | |||
| AK295581.1 | - | 8 | 848 | |||
| AK316042.1 | - | 8 | 841 | |||
| BC032485.1 | - | 9 | 955 | 5221203 | ||
| CR456342.1 | - | 8 | 807 | |||

AIFM3
Genomic Map