siRNA Details
| Gene Symbol: THAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317791.1 | NP_001304720.1 | 4 | 1157 | ||
| NM_144721.5 | NP_653322.1 | 5 | 1283 | |||
| XM_005262772.2 | XP_005262829.1 | 4 | 1178 | |||
| XM_011531666.2 | XP_011529968.1 | 5 | 1720 | |||
| XM_011531667.2 | XP_011529969.1 | 5 | 1243 | |||
| XM_017007800.1 | XP_016863289.1 | 4 | 1323 | |||
| XM_017007801.1 | XP_016863290.1 | 5 | 815 | |||
| GenBank mRNA | AL833350.1 | - | 5 | 1243 | ||
| BC022989.2 | - | 4 | 1197 | 5113206 | ||
| BX647579.1 | - | 4 | 1117 | |||

THAP6
Genomic Map