siRNA Details
| Gene Symbol: ZNF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308123.1 | NP_001295052.1 | 3 | 675 | ||
| NM_006965.3 | NP_008896.2 | 3 | 685 | |||
| XM_005258341.3 | XP_005258398.1 | 3 | 672 | |||
| GenBank mRNA | AF038964.1 | - | 3 | 629 | ||
| AF542097.1 | - | 2 | 506 | |||
| AK291246.1 | - | 3 | 664 | |||
| BC003566.1 | - | 3 | 635 | 3537160 | ||
| BC016801.1 | - | 3 | 656 | 4072599 | ||
| BT006658.1 | - | 2 | 494 | |||
| CR450328.1 | - | 2 | 494 | |||
| U68536.1 | - | 3 | 658 | |||

ZNF24
Genomic Map