siRNA Details
Gene Symbol: ALG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305242.1 | NP_001292171.1 | 5 | 913 | ||
NM_144988.3 | NP_659425.1 | 4 | 876 | |||
NR_131032.1 | 3 | 724 | ||||
GenBank mRNA | AK289395.1 | - | 4 | 876 | ||
BC011706.2 | - | 4 | 852 | 3689162 |
