siRNA Details
| Gene Symbol: LINC00161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026552.1 | 1 | 203 | |||
| GenBank mRNA | AY063458.1 | - | 1 | 203 | ||
| BC101414.1 | - | 1 | 203 | |||
| BC101416.1 | - | 1 | 203 | |||
| BC101419.1 | - | 1 | 203 | |||
| BC101420.1 | - | 1 | 203 | |||

LINC00161
Genomic Map