siRNA Details
Gene Symbol: TUBA3FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003608.1 | 3 | 1396 | |||
GenBank mRNA | AK057257.1 | - | 3 | 1348 | ||
BC009991.2 | - | 3 | 1250 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TUBA3FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003608.1 | 3 | 1396 | |||
GenBank mRNA | AK057257.1 | - | 3 | 1348 | ||
BC009991.2 | - | 3 | 1250 |