siRNA Details
| Gene Symbol: ZBTB47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145166.3 | NP_660149.2 | 6 | 3512 | ||
| GenBank mRNA | AB033016.1 | - | 3 | 1514 | ||
| AK127065.1 | - | 7 | 3206 | |||
| AL133062.1 | - | 5 | 2206 | |||
| BC021855.1 | - | 5 | 2169 | 4943271 | ||

ZBTB47
Genomic Map