siRNA Details
| Gene Symbol: DIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145173.3 | NP_660156.1 | 2 | 742 | ||
| XM_011527713.2 | XP_011526015.1 | 2 | 907 | |||
| GenBank mRNA | AB076888.1 | - | 1 | 683 | ||
| AK124445.1 | - | 1 | 53 | |||
| AY056037.1 | - | 1 | 584 | |||
| AY059641.1 | - | 1 | 584 | |||
| AY180973.1 | - | 1 | 584 | |||
| BC030660.1 | - | 2 | 721 | 4814337 | ||

DIRAS1
Genomic Map