siRNA Details
| Gene Symbol: SPATA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145207.2 | NP_660208.2 | 1, 2 | 243 | ||
| XM_011531679.2 | XP_011529981.1 | 1, 2 | 198 | |||
| XM_017007825.1 | XP_016863314.1 | 1, 2 | 239 | |||
| XM_017007826.1 | XP_016863315.1 | 1, 2 | 239 | |||
| XM_017007827.1 | XP_016863316.1 | 1, 2 | 239 | |||
| XM_017007830.1 | XP_016863319.1 | 1, 2 | 239 | |||
| GenBank mRNA | AF361489.1 | - | 1, 2 | 239 | ||
| AF479656.1 | - | 1, 2 | 213 | |||
| AK091384.1 | - | 1, 2 | 198 | |||
| AL832175.1 | - | 1, 2 | 236 | |||

SPATA5
Genomic Map