siRNA Details
| Gene Symbol: MBD3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145208.2 | NP_660209.2 | 1 | 204 | ||
| GenBank mRNA | AB451462.1 | - | 1 | 118 | ||
| AY038022.1 | - | 1 | 141 | |||
| BC112058.1 | - | 1 | 204 | 8327526 | ||
| BC112064.1 | - | 1 | 204 | 8327532 | ||

MBD3L1
Genomic Map