siRNA Details
| Gene Symbol: EVI5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159944.2 | NP_001153416.1 | 2 | 359 | ||
| NM_145245.4 | NP_660288.1 | 3 | 470 | |||
| XM_005272458.4 | XP_005272515.1 | 3 | 464 | |||
| XM_017026239.1 | XP_016881728.1 | 3 | 456 | |||
| GenBank mRNA | AB449874.1 | - | 2 | 316 | ||
| BC014111.2 | - | 3 | 388 | 4542709 | ||
| BI755965.1 | - | 3 | 425 | |||
| BI913004.1 | - | 3 | 469 | |||

EVI5L
Genomic Map