siRNA Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145248.4 | NP_660291.2 | 1, 2 | 151 | ||
| XM_011537466.1 | XP_011535768.1 | 1, 2 | 176 | |||
| XM_011537468.2 | XP_011535770.1 | 2, 3 | 237 | |||
| XM_011537472.2 | XP_011535774.1 | 1, 2 | 186 | |||
| XM_017020392.1 | XP_016875881.1 | 2, 3 | 252 | |||
| GenBank mRNA | AK130866.1 | - | 1, 2 | 151 | ||
| BC016750.2 | - | 1, 2 | 133 | 4837062 | ||

SPACA7
Genomic Map