siRNA Details
| Gene Symbol: IFI27L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145249.2 | NP_660292.1 | 5 | 654 | ||
| NM_206949.2 | NP_996832.1 | 5 | 608 | |||
| XM_005267314.4 | XP_005267371.1 | 4 | 3199 | |||
| XM_011536405.2 | XP_011534707.1 | 5 | 624 | |||
| XM_011536406.1 | XP_011534708.1 | 6 | 756 | |||
| XM_011536407.1 | XP_011534709.1 | 6 | 759 | |||
| XM_017020961.1 | XP_016876450.1 | 5 | 505 | |||
| XM_017020962.1 | XP_016876451.1 | 4 | 474 | |||
| XM_017020963.1 | XP_016876452.1 | 5 | 591 | |||
| GenBank mRNA | BC015423.1 | - | 5 | 618 | 4398045 | |

IFI27L1
Genomic Map