siRNA Details
| Gene Symbol: SPATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145026.3 | NP_659463.1 | 2, 3 | 240 | ||
| GenBank mRNA | AK058171.1 | - | 1, 2 | 486 | ||
| BC100956.2 | - | 1, 2 | 295 | 40004668 | ||
| BC100957.2 | - | 1, 2 | 295 | 40004669 | ||
| BC100958.2 | - | 1, 2 | 295 | 40004672 | ||
| BC110065.1 | - | 1, 2 | 295 | 40004676 | ||
| BC111414.1 | - | 1, 2 | 295 | 40004677 | ||

SPATS1
Genomic Map