siRNA Details
| Gene Symbol: CCDC151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302453.1 | NP_001289382.1 | 6 | 619 | ||
| NM_001302454.1 | NP_001289383.1 | 4 | 733 | |||
| NM_145045.4 | NP_659482.3 | 6 | 913 | |||
| XM_017026241.1 | XP_016881730.1 | 6 | 911 | |||
| GenBank mRNA | AK056363.1 | - | 6 | 834 | ||
| AK294355.1 | - | 8 | 813 | |||
| AK302113.1 | - | 6 | 620 | |||
| AK308072.1 | - | 4 | 695 | |||
| BC014252.2 | - | 5 | 470 | |||
| BC141828.1 | - | 6 | 845 | 40148425 | ||
| BC142637.1 | - | 6 | 858 | 40147788 | ||
| BC144421.1 | - | 4 | 392 | |||

CCDC151
Genomic Map