siRNA Details
| Gene Symbol: RNF183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145051.3 | NP_659488.2 | 2 | 829 | ||
| XM_006716952.2 | XP_006717015.1 | 4 | 1810 | |||
| XM_011518218.1 | XP_011516520.1 | 4 | 2422 | |||
| XM_011518219.1 | XP_011516521.1 | 3 | 3157 | |||
| XM_011518220.1 | XP_011516522.1 | 4 | 2720 | |||
| XM_017014268.1 | XP_016869757.1 | 3 | 4973 | |||
| GenBank mRNA | AK055759.1 | - | 1 | 2310 | ||
| AK091940.1 | - | 4 | 1807 | |||
| BC013036.2 | - | 2 | 829 | 3535955 | ||

RNF183
Genomic Map