siRNA Details
| Gene Symbol: ZNF625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145233.3 | NP_660276.2 | 4 | 1277 | ||
| NR_037801.1 | 4 | 1273 | ||||
| GenBank mRNA | BC007868.2 | - | 4 | 1256 | 4303049 | |
| BC101589.1 | - | 3 | 1005 | 8069095 | ||
| BC101591.1 | - | 3 | 1005 | 8069097 | ||
| BC143563.1 | - | 3 | 1005 | 9052074 | ||
| Gene Symbol: ZNF625-ZNF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK292455.1 | - | 4 | 1273 | ||

ZNF625
Genomic Map