siRNA Details
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012202.4 | NP_036334.1 | 3 | 879 | ||
| XM_006718500.2 | XP_006718563.1 | 4 | 866 | |||
| GenBank mRNA | AF075042.1 | - | 2 | 542 | ||
| AF087900.1 | - | 3 | 817 | |||
| AF092129.1 | - | 3 | 816 | |||
| BC015563.2 | - | 3 | 760 | 4124714 | ||

GNG3
Genomic Map