siRNA Details
| Gene Symbol: DDIT4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145244.3 | NP_660287.1 | 3 | 403 | ||
| GenBank mRNA | AK094236.1 | - | 3 | 373 | ||
| AK312926.1 | - | 3 | 373 | |||
| AL832277.1 | - | 3 | 355 | |||
| BC013592.2 | - | 3 | 361 | 3877854 | ||
| DB208869.1 | - | 3 | 403 | |||

DDIT4L
Genomic Map