siRNA Details
| Gene Symbol: EVI5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159944.2 | NP_001153416.1 | 2, 3 | 363 | ||
| NM_145245.4 | NP_660288.1 | 3, 4 | 474 | |||
| XM_005272458.4 | XP_005272515.1 | 3, 4 | 468 | |||
| XM_017026239.1 | XP_016881728.1 | 3, 4 | 460 | |||
| GenBank mRNA | AB449874.1 | - | 2, 3 | 320 | ||
| BC014111.2 | - | 3, 4 | 392 | 4542709 | ||
| BI755965.1 | - | 3, 4 | 429 | |||
| BI913004.1 | - | 3, 4 | 473 | |||

EVI5L
Genomic Map