siRNA Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130866.1 | - | 3 | 225 | ||
| BC016750.2 | - | 3 | 207 | 4837062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPACA7| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130866.1 | - | 3 | 225 | ||
| BC016750.2 | - | 3 | 207 | 4837062 | ||