siRNA Details
| Gene Symbol: C10orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 1499 | ||
| XM_017015895.1 | XP_016871384.1 | 3 | 630 | |||
| GenBank mRNA | AK055129.1 | - | 2 | 1477 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf25| Gene Symbol: C10orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039380.3 | NP_001034469.2 | 2 | 1499 | ||
| XM_017015895.1 | XP_016871384.1 | 3 | 630 | |||
| GenBank mRNA | AK055129.1 | - | 2 | 1477 | ||