siRNA Details
| Gene Symbol: UBLCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145049.3 | NP_659486.2 | 11 | 1283 | ||
| GenBank mRNA | AK057996.1 | - | 11 | 1074 | ||
| AY444562.1 | - | 11 | 1095 | |||
| BC013425.2 | - | 11 | 1095 | 3894780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBLCP1| Gene Symbol: UBLCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145049.3 | NP_659486.2 | 11 | 1283 | ||
| GenBank mRNA | AK057996.1 | - | 11 | 1074 | ||
| AY444562.1 | - | 11 | 1095 | |||
| BC013425.2 | - | 11 | 1095 | 3894780 | ||