siRNA Details
| Gene Symbol: DIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145173.3 | NP_660156.1 | 2 | 2454 | ||
| XM_011527713.2 | XP_011526015.1 | 2 | 2619 | |||
| GenBank mRNA | AK124445.1 | - | 1 | 1765 | ||
| AK124672.1 | - | 1 | 1218 | |||
| AL833958.1 | - | 1 | 1243 | |||
| BC030660.1 | - | 2 | 2433 | 4814337 | ||

DIRAS1
Genomic Map