siRNA Details
| Gene Symbol: SLC2A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145176.2 | NP_660159.1 | 5 | 2202 | ||
| GenBank mRNA | AK056554.1 | - | 4 | 941 | ||
| AK122628.1 | - | 5 | 2194 | |||
| AK226098.1 | - | 5 | 2073 | |||
| AL833602.1 | - | 5 | 2186 | |||
| AY046419.1 | - | 5 | 2164 | |||
| BC070149.1 | - | 5 | 2136 | 30371087 | ||
| BX648889.1 | - | 6 | 1337 | |||

SLC2A12
Genomic Map