siRNA Details
| Gene Symbol: GMEB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012384.4 | NP_036516.1 | 5 | 595 | ||
| XM_005260202.4 | XP_005260259.1 | 5 | 806 | |||
| XM_006723776.2 | XP_006723839.1 | 3 | 247 | |||
| XM_011528779.2 | XP_011527081.1 | 3 | 311 | |||
| GenBank mRNA | AB033095.1 | - | 5 | 472 | ||
| AF173867.1 | - | 4 | 871 | |||
| AL133646.1 | - | 3 | 319 | |||
| BC036305.1 | - | 5 | 522 | 5166938 | ||
| BQ954281.1 | - | 4 | 297 | |||

GMEB2
Genomic Map