siRNA Details
| Gene Symbol: SPATA18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297608.1 | NP_001284537.1 | 1, 2 | 460 | ||
| NM_145263.3 | NP_660306.1 | 1, 2 | 460 | |||
| NR_123728.1 | 1, 2 | 460 | ||||
| XM_005265716.1 | XP_005265773.1 | 1, 2 | 375 | |||
| XM_005265717.2 | XP_005265774.1 | 1, 2 | 375 | |||
| XM_011534369.1 | XP_011532671.1 | 1, 2 | 375 | |||
| XM_011534370.1 | XP_011532672.1 | 1, 2 | 375 | |||
| GenBank mRNA | AB465501.1 | - | 1, 2 | 86 | ||
| AB465502.1 | - | 1, 2 | 86 | |||
| AK057468.1 | - | 1, 2 | 372 | |||
| AK304386.1 | - | 1, 2 | 375 | |||
| AK314799.1 | - | 1, 2 | 330 | |||
| BC025396.1 | - | 1, 2 | 367 | 4821894 | ||
| DC404003.1 | - | 1, 2 | 460 | |||

SPATA18
Genomic Map