siRNA Details
| Gene Symbol: CD300LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168322.1 | NP_001161794.1 | 2 | 146 | ||
| NM_001168323.1 | NP_001161795.1 | 2 | 146 | |||
| NM_001168324.1 | NP_001161796.1 | 2 | 146 | |||
| NM_145273.3 | NP_660316.2 | 2 | 146 | |||
| XM_005257075.2 | XP_005257132.1 | 2 | 517 | |||
| XM_005257076.2 | XP_005257133.1 | 2 | 517 | |||
| XM_006721721.2 | XP_006721784.1 | 2 | 517 | |||
| XM_006721722.2 | XP_006721785.1 | 2 | 517 | |||
| XM_011524384.2 | XP_011522686.1 | 2 | 517 | |||
| XM_017024235.1 | XP_016879724.1 | 2 | 517 | |||
| GenBank mRNA | AF427619.1 | - | 2 | 138 | ||
| AF427620.1 | - | 2 | 138 | |||
| AK298112.1 | - | 2 | 148 | |||
| AK309938.1 | - | 2 | 147 | |||
| AY358364.1 | - | 2 | 101 | |||
| BC025395.2 | - | 2 | 118 | 4827737 | ||
| DC380538.1 | - | 2 | 520 | |||

CD300LG
Genomic Map