siRNA Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145658.3 | NP_663633.1 | 2 | 319 | ||
| GenBank mRNA | AF275321.1 | - | 2 | 223 | ||
| AY358983.1 | - | 2 | 302 | |||
| BC017998.1 | - | 2 | 293 | 4252566 | ||
| BG720873.1 | - | 2 | 315 | |||
| BM564110.1 | - | 2 | 318 | |||

SPESP1
Genomic Map