siRNA Details
| Gene Symbol: ABT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013375.3 | NP_037507.1 | 3 | 1179 | ||
| GenBank mRNA | BC048812.1 | - | 3 | 1177 | 5574755 | |
| BC066313.1 | - | 3 | 1169 | 5272771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABT1| Gene Symbol: ABT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013375.3 | NP_037507.1 | 3 | 1179 | ||
| GenBank mRNA | BC048812.1 | - | 3 | 1177 | 5574755 | |
| BC066313.1 | - | 3 | 1169 | 5272771 | ||