siRNA Details
| Gene Symbol: MTPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145808.3 | NP_665807.1 | 1, 2 | 436 | ||
| GenBank mRNA | AF106075.1 | - | 1, 2 | 285 | ||
| AK056225.1 | - | 1, 2 | 339 | |||
| AL834231.1 | - | 1, 2 | 308 | |||
| BC028093.1 | - | 1, 2 | 334 | 3876632 | ||
| DA325130.1 | - | 1, 2 | 436 | |||
| Gene Symbol: LUZP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128619.2 | NP_001122091.2 | 1, 2 | 436 | ||
| GenBank mRNA | DA325904.1 | - | 1, 2 | 358 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008098, AK145755, AK146473, BC054811, AK152317, and others.

MTPN
Genomic Map