siRNA Details
| Gene Symbol: HOXC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014212.3 | NP_055027.1 | 1 | 733 | ||
| GenBank mRNA | AJ000041.1 | - | 1 | 661 | ||
| AK291946.1 | - | 1 | 731 | |||
| BC001543.1 | - | 1 | 714 | 3462682 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXC11| Gene Symbol: HOXC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014212.3 | NP_055027.1 | 1 | 733 | ||
| GenBank mRNA | AJ000041.1 | - | 1 | 661 | ||
| AK291946.1 | - | 1 | 731 | |||
| BC001543.1 | - | 1 | 714 | 3462682 | ||