siRNA Details
| Gene Symbol: EVC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166136.1 | NP_001159608.1 | 4 | 978 | ||
| NM_147127.4 | NP_667338.3 | 4 | 540 | |||
| XM_017007735.1 | XP_016863224.1 | 4 | 551 | |||
| XM_017007736.1 | XP_016863225.1 | 5 | 529 | |||
| XM_017007737.1 | XP_016863226.1 | 5 | 416 | |||
| XM_017007738.1 | XP_016863227.1 | 4 | 551 | |||
| XM_017007739.1 | XP_016863228.1 | 5 | 582 | |||
| GenBank mRNA | AB083067.1 | - | 3 | 246 | ||
| AY152402.1 | - | 4 | 975 | |||
| AY152403.1 | - | 4 | 975 | |||
| AY185210.1 | - | 4 | 540 | |||
| BC143836.1 | - | 5 | 587 | |||

EVC2
Genomic Map