siRNA Details
| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145276.2 | NP_660319.1 | 4 | 2676 | ||
| GenBank mRNA | AK092116.1 | - | 6 | 1974 | ||
| BC022523.1 | - | 4 | 2644 | 4794529 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF563| Gene Symbol: ZNF563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145276.2 | NP_660319.1 | 4 | 2676 | ||
| GenBank mRNA | AK092116.1 | - | 6 | 1974 | ||
| BC022523.1 | - | 4 | 2644 | 4794529 | ||