siRNA Details
| Gene Symbol: UBXN11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145345.2 | NP_663320.2 | 5 | 549 | ||
| NM_183008.2 | NP_892120.2 | 6 | 554 | |||
| GenBank mRNA | AF520989.1 | - | 4 | 258 | ||
| AF521017.1 | - | 4 | 259 | |||
| AF521018.1 | - | 4 | 247 | |||
| AK074358.1 | - | 1 | 46 | |||
| AK090792.1 | - | 2 | 124 | |||
| AK093258.1 | - | 6 | 498 | |||
| AK097368.1 | - | 5 | 647 | |||
| AK291503.1 | - | 4 | 254 | |||
| AK296179.1 | - | 3 | 156 | |||
| AY007163.1 | - | 3 | 140 | |||
| BC038106.1 | - | 5 | 366 | 5269682 | ||
| BX647774.1 | - | 5 | 362 | |||
| BX648412.1 | - | 4 | 263 | |||
| CR749413.1 | - | 5 | 360 | |||
| DA421971.1 | - | 5 | 361 | |||

UBXN11
Genomic Map