siRNA Details
| Gene Symbol: GTF3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035521.2 | NP_001030598.1 | 19 | 3394 | ||
| NM_001318909.1 | NP_001305838.1 | 19 | 3504 | |||
| NM_001521.3 | NP_001512.1 | 20 | 3470 | |||
| XM_005264272.4 | XP_005264329.1 | 20 | 3678 | |||
| XM_011532801.2 | XP_011531103.1 | 20 | 3817 | |||
| GenBank mRNA | AF054988.1 | - | 7 | 1219 | ||
| BC020981.1 | - | 18 | 3103 | 3849379 | ||
| BX647092.1 | - | 19 | 3504 | |||
| D13636.1 | - | 18 | 3113 | |||

GTF3C2
Genomic Map