siRNA Details
| Gene Symbol: ABTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145804.2 | NP_665803.2 | 9 | 2380 | ||
| GenBank mRNA | AK091560.1 | - | 6 | 598 | ||
| AK095632.1 | - | 9 | 1627 | |||
| AK291744.1 | - | 9 | 2381 | |||
| BC093968.1 | - | 9 | 1492 | 7939813 | ||
| BX640841.1 | - | 1 | 81 | |||

ABTB2
Genomic Map