siRNA Details
| Gene Symbol: CCSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145257.4 | NP_660300.3 | 4 | 988 | ||
| GenBank mRNA | AK123465.1 | - | 3 | 777 | ||
| AK291437.1 | - | 3 | 949 | |||
| BC015419.1 | - | 3 | 962 | 4396825 | ||
| BC060777.1 | - | 4 | 1893 | |||
| BC071609.1 | - | 4 | 1034 | 5264741 | ||

CCSAP
Genomic Map