siRNA Details
| Gene Symbol: TMEM99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195386.1 | NP_001182315.1 | 3 | 860 | ||
| NM_001195387.1 | NP_001182316.1 | 3 | 844 | |||
| NM_145274.3 | NP_660317.2 | 3 | 852 | |||
| XM_005257089.4 | XP_005257146.1 | 4 | 1049 | |||
| XM_017024253.1 | XP_016879742.1 | 4 | 1017 | |||
| GenBank mRNA | AK097454.1 | - | 3 | 793 | ||
| AK298616.1 | - | 3 | 795 | |||
| BC015365.1 | - | 3 | 831 | 3900532 | ||

TMEM99
Genomic Map