siRNA Details
| Gene Symbol: STOML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144033.1 | NP_001137505.1 | 8 | 1254 | ||
| NM_145286.2 | NP_660329.1 | 7 | 1170 | |||
| XM_017020400.1 | XP_016875889.1 | 5 | 878 | |||
| GenBank mRNA | BC025760.1 | - | 7 | 1157 | 5204252 | |

STOML3
Genomic Map