siRNA Details
| Gene Symbol: ZNF519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145287.3 | NP_660330.2 | 3 | 492 | ||
| XM_011525605.2 | XP_011523907.1 | 2 | 652 | |||
| XM_017025562.1 | XP_016881051.1 | 3 | 496 | |||
| XM_017025563.1 | XP_016881052.1 | 3 | 496 | |||
| XM_017025564.1 | XP_016881053.1 | 3 | 496 | |||
| XM_017025565.1 | XP_016881054.1 | 3 | 496 | |||
| XM_017025566.1 | XP_016881055.1 | 2 | 353 | |||
| GenBank mRNA | AB209417.1 | - | 1 | 179 | ||
| BC024227.1 | - | 3 | 436 | 4806717 | ||

ZNF519
Genomic Map